Alfa Aesar™ Copper molybdenum oxide, 98%

Paint pigment, corrosion inhibitor

Manufacturer: Alfa Aesar™ 8993622

Quantity 100g

 View more versions of this product

Catalog No. AA8993622

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 13767-34-5
    Molecular Formula CuMoO4
    Formula Weight 223.49
    MDL Number MFCD00016057
    Synonym Copper molybdate
    Chemical Name or Material Copper molybdenum oxide

    Specifications

    Formula Weight 223.49
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    MDL Number MFCD00016057
    Synonym Copper molybdate
    Density 3.4g/mL
    Physical Form Powder
    Molecular Formula CuMoO4
    Solubility Information Insoluble in water
    Health Hazard 2 GHS H Statement
    H373-H319-H335
    May cause damage to organs through prolonged or repeated exposure.
    Causes serious eye irritation.
    May cause respiratory irritation.
    CAS 13767-34-5
    Melting Point ≈500°C
    Quantity 100g
    Health Hazard 1 Warning
    Chemical Name or Material Copper molybdenum oxide
    Assay 98%

    GHS H Statement
    H373-H319-H335
    May cause damage to organs through prolonged or repeated exposure.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit