Alfa Aesar™ Copper, Oil based standard solution, Specpure™, Cu 1000µg/g

Manufacturer: Alfa Aesar™ 4727618

Quantity 50g

 View more versions of this product

Catalog No. AA4727618

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00010965
    Chemical Name or Material Copper, Oil based standard solution
    Grade Specpure™

    Specifications

    Quantity 50g
    Physical Form Liquid
    Concentration Cu 1000µg/g
    MDL Number MFCD00010965
    Chemical Name or Material Copper, Oil based standard solution
    Grade Specpure™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit