Copper Plate, Oxygen-Free High Conductivity (OFHC), alloy 101, 6.35mm (0.25in.) thick, , Alfa Aesar™

Manufacturer: Alfa Aesar 45190HB

Quantity 300 x 300mm

 View more versions of this product

Catalog No. AA45190HB

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Plate, Oxygen-Free High Conductivity (Ofhc), Alloy 101, 6.35mm (0.25 in.) thick
    MDL Number MFCD00010965
    Chemical Name or Material Copper

    Specifications

    Physical Form Plate, Oxygen-Free High Conductivity (Ofhc), Alloy 101, 6.35mm (0.25 in.) thick
    Quantity 300 x 300mm
    Color Copper colored
    MDL Number MFCD00010965
    Odor Odorless
    Appearance Copper colored plate
    Chemical Name or Material Copper

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit