Alfa Aesar™ Copper powder, 5% in graphite

Manufacturer: Alfa Aesar™ 8968822

Quantity 100g

 View more versions of this product

Catalog No. AA8968822

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00010965
    UN Number UN1325
    Chemical Name or Material Copper powder, Graphimet Cu-5

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    UN Number UN1325
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    TSCA TSCA
    MDL Number MFCD00010965
    Health Hazard 2 GHS H Statement
    H228-H319-H335
    Flammable solid.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Concentration 5% in graphite
    Quantity 100g
    Chemical Name or Material Copper powder, Graphimet Cu-5

    GHS H Statement
    H228-H319-H335
    Flammable solid.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit