Alfa Aesar™ Copper powder, semispherical, -50+70 mesh, 99% (metals basis)

Manufacturer: Alfa Aesar™ 4503036

Quantity 500g

 View more versions of this product

Catalog No. AA4503036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-50-8
    MDL Number MFCD00010965
    UN Number UN3077
    Chemical Name or Material Copper powder

    Specifications

    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 7440-50-8
    Mesh Size −50+70 Mesh
    Health Hazard 2 GHS H Statement
    H319-H335
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    MDL Number MFCD00010965
    Odor Odorless
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Quantity 500g
    Physical Form Semispherical
    UN Number UN3077
    Health Hazard 1 Warning
    Chemical Name or Material Copper powder
    Assay 99%, (metals basis)

    GHS H Statement
    H319-H335
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit