Alfa Aesar™ Copper powder, spherical, -170+400 mesh, 99.9% (metals basis), Oxygen <1000ppm

Manufacturer: Alfa Aesar™ 43385A3

Quantity 2kg

 View more versions of this product

Catalog No. AA43385A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-50-8
    MDL Number MFCD00010965
    UN Number UN3089
    Chemical Name or Material Copper powder, Oxygen <1000ppm

    Specifications

    Physical Form Sperical Powder
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color Copper
    CAS 7440-50-8
    Health Hazard 2 GHS H Statement
    H228-H319-H335
    Flammable solid.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 2kg
    TSCA TSCA
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    Mesh Size −170+400 Mesh
    Odor Odorless
    RTECS Number GL5325000
    MDL Number MFCD00010965
    UN Number UN3089
    Health Hazard 1 Warning
    Chemical Name or Material Copper powder, Oxygen <1000ppm
    Assay 99.9%, (metals basis)

    GHS H Statement
    H228-H319-H335
    Flammable solid.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit