Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Copper rod, Oxygen-Free High Conductivity (OFHC), alloy 101, 9.53mm (0.375 in.) dia. 183cm

Manufacturer:  Alfa Aesar™ 45189AEX

Quantity 183cm

 View more versions of this product

Catalog No. AA45189AEX


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD00010965
Copper rod, Oxygen-Free High Conductivity (OFHC), alloy 101, 9.53mm (0.375 in.) dia.

Specifications

183cm
MFCD00010965
Odorless
Copper rod, Oxygen-Free High Conductivity (OFHC), alloy 101, 9.53mm (0.375 in.) dia.

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit