Alfa Aesar™ Copper Silicon bar, 3.8x3.8x30cm

Manufacturer: Alfa Aesar™ 43767KG

Quantity 1 piece

 View more versions of this product

Catalog No. AA43767KG

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD02684526
    Synonym Everdur™ Silicon Bronze
    Chemical Name or Material Copper Silicon bar, 3.8x3.8x30cm

    Specifications

    Synonym Everdur™ Silicon Bronze
    Avg. Mol. Wt. or Mol. Wt. Range Cu:Si; 97:3 wt%
    TSCA TSCA
    MDL Number MFCD02684526
    Weight ≈3.7kg/bar
    Quantity 1 piece
    Chemical Name or Material Copper Silicon bar, 3.8x3.8x30cm

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit