Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Plastic bottle
7440-50-8, 7697-37-2, 7732-18-5
Cu
63.546
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
CHEBI:30052
copper
[Cu]
7440-50-8, 7697-37-2, 7732-18-5
Cu
63.546
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
CHEBI:30052
copper
[Cu]
For atomic adsorption spectroscopy
Solubility Solubility in water: soluble.
Concentration 997 to 1003 μg/mL of Cu(2+) in 2% HNO3 (20°C)
Fieser 16,95; 16,181
Formula Weight 63.55
Physical Form Solution
Grade AAS
Packaging Plastic bottle
Density 8.92
Color Blue
Quantity 100mL
Chemical Name or Material Copper standard solution, 1 mg/ml Cu in 2% HNO3
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May be corrosive to metals.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning