Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Sulfate, 2 g/L Solution, For Morpholine, ASTM, Spectrum™

Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
Grade Chemical Solution
CAS 7758-99-8,7732-18-5
Packaging Poly Bottle
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.