Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper Sulfate, For Nitrogen (Nitrate), Certified, 2.0% (w/v) ±0.1%, LabChem™
$45.70 - $163.00
Specifications
Color | Clear, blue |
---|---|
Format | Liquid |
Packaging | Poly Bottle |
Storage Requirements | Room Temperature |
Type | Copper Sulfate |
Specifications
Clear, blue | |
Poly Bottle | |
Copper Sulfate |
Liquid | |
Room Temperature |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title