Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Sulfate-Hydrochloric Acid, Solution, Spectrum™

Click to view available options
Quantity:
4 L
Packaging:
Poly Bottle
Grade Chemical Solution
CAS 7647-01-0,7732-18-5,7758-99-8
Packaging Poly Bottle
Quantity 4 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.