Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Sulfate, Pentahydrate, Granular, FCC, 98-102%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7758-99-8
CuH10O9S
249.68
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper(2+) pentahydrate sulfate
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
7758-99-8
CuH10O9S
249.68
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper(2+) pentahydrate sulfate
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

C1431, 7758-99-8, CuSO4•5H2O

Spectrum™ Copper Sulfate, Pentahydrate, Granular, FCC is used as a fungicide, algaecide and feed supplement. Find a high quality inventory of Copper Copper Sulfate, Pentahydrate, Granular, FCC. Approved by the Food Chemical Codex, it is used as a fungicide, algaecide, and feed supplement, and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
UN Number UN3077
Percent Purity 98-102%
Lead (Pb) 4 mg/kg
Packaging Poly Pail
Quantity 2.5 kg