Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Sulfate Solution, ASTM A380/A967, Reagents

Catalog No. p-200021686
Click to view available options
Quantity:
500 mL
4 L

Copper Sulfate Solution, ASTM A380/A967, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Copper Sulfate Solution, ASTM A380/A967
Boiling Point 100°C
CAS 7732-18-5, 7758-99-8
Color Blue
pH <1
Physical Form Liquid
Quantity 4 L
Molecular Formula CuSO4/H2O/H2SO4
Synonym Copper(II) Sulfate
Percent Purity 1.6%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.01
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.