Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper Sulfate Solution (For AOAC Method 930.28), Fisher Chemical™
Click to view available options
Quantity:
250 mL
500 mL
Packaging:
Natural Poly Bottle
Specifications
CAS Min % | 92.0, 6.35 |
CAS Max % | 95.0, 6.61 |
Packaging | Natural Poly Bottle |
Methods | For AOAC Method 930.28 |
Quantity | 250 mL |
Chemical Name or Material | Copper Sulfate Solution |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction