Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Sulfate Solution, Specific Gravity 1.186 at 18.3°C, Spectrum™

Click to view available options
Quantity:
1 L
4 L
Packaging:
Poly Bottle
7758-99-8,7732-18-5
7758-99-8,7732-18-5
Grade Chemical Solution
Packaging Poly Bottle
Specific Gravity 1.183 - 1.189
pH 3
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.