Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Copper Sulfate Solution, Spectrum™ Chemical
Specifications
CAS | 7758-99-8, 7732-18-5 |
---|---|
Grade | Chemical Solution |
Packaging | Poly Bottle |
Specific Gravity | 1.052 - 1.054 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
18-609-948
|
Spectrum Chemical Manufacturing Corporation
C2071LT |
1 L | Poly Bottle | N/A | N/A | N/A | |||
Specifications
7758-99-8, 7732-18-5 | |
Chemical Solution | |
Poly Bottle | |
1.052 - 1.054 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title