Alfa Aesar™ Copper Thinfoil, 0.015mm (0.0006 in.) thick, 99.97% (metals basis)

Manufacturer: Alfa Aesar™ 41504FF

Quantity 25x25mm

 View more versions of this product

Catalog No. AA41504FF

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7440-50-8
    MDL Number MFCD00010965
    Chemical Name or Material Copper Thinfoil, 0.015mm (0.0006 in.) thick, Not certified pinhole free

    Specifications

    MDL Number MFCD00010965
    CAS 7440-50-8
    Odor Odorless
    Weight ≈0.084g/25x25mm
    Quantity 25x25mm
    TSCA TSCA
    Physical Form Foil
    Chemical Name or Material Copper Thinfoil, 0.015mm (0.0006 in.) thick, Not certified pinhole free
    Assay 99.97%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit