Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Copper wire, 0.5mm (0.02 in.) dia., Puratronic™, 99.999% (metals basis), Oxygen free

Click to view available options
Quantity:
50cm
250cm
7440-50-8
Cu
63.546
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer
23978
CHEBI:30052
copper
[Cu]
7440-50-8
Cu
63.546
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer
23978
CHEBI:30052
copper
[Cu]
Quantity 50cm
Weight ≈1.75g/m
Odor Odorless
Physical Form Wire, ≈1.75g/m
Percent Purity 99.999%
Grade Puratronic™
Chemical Name or Material Copper wire, 0.5mm (0.02 in.) dia., Oxygen free
Assay (metals basis)

EINECSNumber : 231-159-6

TSCA : Yes

Recommended Storage : Ambient temperatures