Learn More
COPS2 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB30005
Catalog No. 89-033-293
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human COPS2.
  • High specificity
  • High reactivity
Specifications
Specifications
COPS2
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (2.5 ug/mL) The optimal working dilution should be determined by the end user.
NM_004236
ALIEN|CSN2|SGN2|TRIP15
Rabbit
Protein A purification
RUO
Primary
9318
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human COPS2.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
COPS2
A synthetic peptide corresponding to N-terminus of human COPS2.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)