Learn More
COPS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 155771AP
Catalog No. 50-173-0325
Add to cart
Description
Description
The protein encoded by this gene possesses kinase activity that phosphorylates regulators involved in signal transduction. It phosphorylates I kappa-Balpha, p105, and c-Jun. It acts as a docking site for complex-mediated phosphorylation. The gene is located within the Smith-Magenis syndrome region on chromosome 17.
Specifications
Specifications
COPS3
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88543, Q68FW9, Q9UNS2
COPS3, CSN3, SGN3, Signalosome subunit 3
Rabbit
IgG
150 μL
-20°C
Primary
26572, 287367, 8533
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
COPS3
Liquid
Cops3
COPS3 Fusion Protein Ag7968
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat