Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COPS5 Mouse anti-Human, Mouse, Clone: 2A10.8, Abnova™

Mouse monoclonal antibody raised against full length recombinant mouse Cops5.

Manufacturer:  Abnova Corporation MAB12917

Catalog No. 89-933-986


Add to cart

Description

Description

Specifications

Specifications

COPS5
2A10.8
Mouse monoclonal antibody raised against full length recombinant mouse Cops5.
In PBS, pH 7.4.
AI303502/CSN5/Jab1/Mov34/Sgn5
Mouse
IgG2b
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Murine
Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Liquid
Cops5
Recombinant protein corresponding to full length mouse Cops5.
Protein G purification
RUO
Primary
26754
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit