Learn More
Abnova™ COPS6 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00010980T01
Catalog No. 89-940-218
Add to cart
Description
Description
Specifications
Specifications
Western Blot
COPS6 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 36.2kDa
Transient overexpression cell lysate was tested with anti-COPS6 antibody (Mfr. No. H00010980-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_006833
pCMV-COPS6 full-length
100 μL
Human