Learn More
COPZ1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 204401AP
Catalog No. 50-173-2890
Add to cart
Description
Description
This gene encodes a subunit of the cytoplasmic coatamer protein complex, which is involved in autophagy and intracellular protein trafficking. The coatomer protein complex is comprised of seven subunits and functions as the coat protein of coat protein complex (COP)I-vesicles. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
COPZ1
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P61923, P61924
CGI 120, Coatomer subunit zeta 1, COPZ, COPZ1, HSPC181, Zeta 1 coat protein, Zeta 1 COP, zeta1 COP
Rabbit
IgG
150 μL
-20°C
Primary
22818, 315345, 56447
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
COPZ1
Liquid
Copz1
COPZ1 Fusion Protein Ag14234
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate, Rat