Learn More
Invitrogen™ COQ6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114228
Description
Immunogen sequence: GQGVNMGFGD ISSLAHHLST AAFNGKDLGS VSHLTGYETE RQRHNTALLA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 100%; Rat: 93%; Zebrafish: 77%.
COQ6 (Coenzyme Q10 monooxygenase 6) is also named as CGI-10 and belongs to the ubiH/COQ6 family. It is an evolutionarily conserved flavin-dependent monooxygenase required for biosynthesis of coenzyme Q10, or ubiquinone. It has 3 isoforms with the molecular mass of 51 kDa, 43 kDa and 49 kDa. In inner ear, Coq6 is expressed in spiral ganglion, stria vascularis cells, and spiral ligament. Defects in COQ6 are the cause of coenzyme Q10 deficiency, primary, type 6 (COQ10D6).
Specifications
COQ6 | |
Polyclonal | |
Unconjugated | |
Coq6 | |
5930427M12Rik; AW742344; CGI10; CGI-10; Coenzyme Q10 monooxygenase 6; coenzyme Q6 homolog (yeast); coenzyme Q6 homolog, monooxygenase; coenzyme Q6 monooxygenase; coenzyme Q6, monooxygenase; COQ10D6; Coq6; ubiquinone biosynthesis monooxygenase COQ6; ubiquinone biosynthesis monooxygenase COQ6, mitochondrial | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51004 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y2Z9 | |
Coq6 | |
Immunogen sequence: GQGVNMGFGD ISSLAHHLST AAFNGKDLGS VSHLTGYETE RQRHNTALLA. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.