Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COQ6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP198412
Description
COQ6 Polyclonal specifically detects COQ6 in Human samples. It is validated for Western Blot.Specifications
COQ6 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
NP_872286 | |
COQ6 | |
The immunogen for this antibody is COQ6 - C-terminal region. Peptide sequence GQGVNMGFGDISSLAHHLSTAAFNGKDLGSVSHLTGYETERQRHNTALLA. | |
Affinity Purified | |
RUO | |
51004 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CGI-10, coenzyme Q6 homolog (yeast), coenzyme Q6 homolog, monooxygenase (S. cerevisiae), coenzyme Q6 homolog, monooxygenase (yeast), EC 1.14.13, EC 1.14.13.-, ubiquinone biosynthesis monooxygenase COQ6 | |
Rabbit | |
48 kDa | |
100 μL | |
Primary | |
Bovine: 86%; Guinea pig: 86%; Zebrafish: 77%. | |
Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title