Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ COQ7 Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793925PLATES

 View more versions of this product

Catalog No. NB009824


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Human COQ7.
Specifications

Specifications

Sandwich ELISA
HRP
39.1 pg/mL
10229
RUO
COQ7
Stem Cell Markers
Human
COQ7
1. Capture Antibody: Dilute to a working concentration of 1.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2500 pg/mL is recommended.
Antibody Pairs
This solid phase sandwich ELISA utilizes an antibody specific for Human COQ7.
Sandwich ELISA
  • Standard: Each vial contains 100 ng of recombinant COQ7.
  • Capture Antibody: 0.4 mg/mL of mouse anti-COQ7 monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL rabbit anti-COQ7 polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Storage is content dependent.
    Rabbit/Mouse
    5 Plates
    39.06-2500 pg/mL
    CAT5, CLK1, CLK-1, Coenzyme Q biosynthesis protein 7 homolog, coenzyme Q, 7 (rat, yeast) homolog, coenzyme Q7 homolog, ubiquinone (yeast), COQ7 coenzyme Q, 7 homolog ubiquinone, placental protein KG-20, Timing protein clk-1 homolog, ubiquinone biosynthesis protein COQ7 homolog
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit