Learn More
Allcells LLCSupplier Diversity Partner Cord Blood, Cryopreserved, CD34+ Stem/Progenitor Cells, Positively Selected, Single 0.5M DFS Item
Manufacturer:  Allcells LLCSupplier Diversity Partner CBCRCD34+SINGLEPS0.5M
AllCells CB-CD34+ Stem/Progenitor cells are positively isolated using a direct immunomagnetic CD34 MicroBead labeling system.CB-CD34+ Cells/Mixed indicates that multiple donors are pooled to obtain the cord blood CD34+ cells.
Catalog No. 50-489-382