Learn More
SMARTLOGICS LLC Core Gloves - Medium- 9 inch Cuff - Nitrile, 3-4 Mil, Laboratory Gloves, Latex Free, Allergy Free, Accelerator-Free for Sensitive Skin, Powder-Free with Textured Grip, Ambidextrous, Blue, Case of 1001
Supplier:  SMARTLOGICS LLC CGNMM 01
Encompass_Preferred
Gloves, Safety, Nitrile, Laboratory Grade, Core Gloves, Regular 9.5 inch Cuff, MediumDependable 3-4 Mil Thickness: robust and reliable barrier against chemicals and solvents while preserving tactile sensitivity, Enhanced Grip via Textured Fingertips.Ambidextrous and Non-Sterile. Advanced Accelerator-Free Formulation: Engineered for professionals with sensitive skin, 100% Free of Natural Rubber Latex and our unique nitrile blend prevents Type I and Type IV chemical allergies, allowing for extended, comfortable wear without irritation during critical tasks.High-Purity Powder-Free Design: preventing downstream contamination, and protecting the integrity of your research, Color: Professional Blue; Optimized for demanding applications in biotechnology, pharmaceutical labs, electronics assembly where precision and purity are paramount. 100 gloves by weight per convenient dispenser box, 10 boxes per case (1000 gloves total), designed for efficient laboratory supply and workflow.
Catalog No. NC3803807