Learn More
Fisherbrand™ Core Vinyl Chair - High Bench Height with Adjustable Arms, Chrome Foot Ring, and Stationary Glides in Grade C Vinyl
Fisherbrand™ Core Vinyl Chairs with stationary glides are height adjustable for 42 in. work surfaces and feature manual back height and depth adjustments, arms, and chrome foot ring making them ideal for educational environments.Offers antifungal Grade C vinyl in a variety of colors.
$253.00
Specifications
Adjustable Yes (Pneumatic)
Material Vinyl
Footrest Yes
With Armrest Yes
With Backrest Yes
View More Specs
Products 6
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S39175 Products for Science Education
View Documents
Orange
Each for $253.00
Add to cart
S39174 Products for Science Education
View Documents
Dark Green
Each for $253.00
Add to cart
S39172 Products for Science Education
View Documents
Red
Each for $253.00
Add to cart
S39173 Products for Science Education
View Documents
Light Blue
Each for $253.00
Add to cart
S39176 Products for Science Education
View Documents
Brown
Each for $253.00
Add to cart
S39177 Products for Science Education
View Documents
Lime Green
Each for $253.00
Add to cart
Description
Description
  • Adjustable from 24-34 inches from the top of the seat to the floor
  • Features manual back and back depth adjustments
  • 2-way adjustable arm rests
  • 18-Inch Chrome adjustable foot ring with knob lock
  • Black, nylon-reinforced fiberglass base
  • Stationary, non-marring glides suitable for hard floors
  • Antifungal/antibacterial Grade C Vinyl.
Specifications
Specifications
Yes (Pneumatic)
Yes
Yes
No
Vinyl
Yes
No