Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Fisherbrand™ Core Vinyl Chair - Medium Bench Height with Black Foot Ring and Stationary Glides in Grade C Vinyl
Fisherbrand™ Core Vinyl Chairs with stationary glides are height adjustable for 36 in. work surfaces and feature manual back height and depth adjustments and black adjustable foot ring making them ideal for educational environments.Offers antifungal Grade C vinyl in a variety of colors.
Specifications
Adjustable Yes (Pneumatic)
Material Vinyl
Footrest Yes
With Armrest No
With Backrest Yes
View More Specs
Products 6
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S39199 Products for Science Education
View Documents
Orange
Each for $198.00
N/A
S39198 Products for Science Education
View Documents
Dark Green
Each for $198.00
N/A
S39196 Products for Science Education
View Documents
Red
Each for $198.00
N/A
S39197 Products for Science Education
View Documents
Light Blue
Each for $198.00
N/A
S39200 Products for Science Education
View Documents
Brown
Each for $198.00
N/A
S39201 Products for Science Education
View Documents
Lime Green
Each for $198.00
N/A
Description
Description
  • Adjustable from 19.5-27.5 inches from the top of the seat to the floor
  • Features manual back and back depth adjustments
  • 18-Inch black nylon adjustable foot ring with knob lock
  • Black, nylon-reinforced fiberglass base
  • Stationary, non-marring glides suitable for hard floors
  • Antifungal/antibacterial Grade C Vinyl
Specifications
Specifications
Yes (Pneumatic)
Yes
Yes
No
Vinyl
No
No