Learn More
Fisherbrand™ Core Vinyl Chair - Medium Bench Height with Round Tube Base, Adjustable Arms, and Stationary Glides in Grade C Vinyl
Fisherbrand™ Core Vinyl Chairs with Round Tube Base are height adjustable for 36 in. work surfaces and feature manual adjustments, fixed foot ring, 2-way adjustable arms, and stationary glides making them ideal for educational environments.Offers antifungal Grade C in a variety of colors.
$238.00
Specifications
Adjustable Yes (Pneumatic)
Material Vinyl
Footrest No
With Armrest Yes
With Backrest Yes
View More Specs
Products 6
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S39181 Products for Science Education
View Documents
Black
Each for $238.00
Add to cart
S39180 Products for Science Education
View Documents
Black
Each for $238.00
Add to cart
S39178 Products for Science Education
View Documents
Black
Each for $238.00
Add to cart
S39179 Products for Science Education
View Documents
Black
Each for $238.00
Add to cart
S39182 Products for Science Education
View Documents
Black
Each for $238.00
Add to cart
S39183 Products for Science Education
View Documents
Black
Each for $238.00
Add to cart
Description
Description
  • Adjustable from 22-30 inches from the top of the seat to the floor
  • Features manual back and back depth adjustments
  • 2-way adjustable arm rests
  • Black, powder-coated steel base with fixed foot ring
  • Stationary, non-marring glides suitable for hard floors
  • Antifungal/antibacterial Grade C Vinyl
Specifications
Specifications
Yes (Pneumatic)
No
Yes
No
Vinyl
Yes
No