Learn More
Fisherbrand™ Core Vinyl Chair - Medium Bench Height with Round Tube Base and Stationary Glides in Grade C Vinyl
Fisherbrand™ Core Vinyl Chairs with Round Tube Base are height adjustable for 36 in. work surfaces and feature manual adjustments, fixed foot ring, and stationary glides making them ideal for educational environments.Offers antifungal Grade C in a variety of colors.
$195.00
Specifications
Adjustable Yes (Pneumatic)
Material Vinyl
Footrest No
With Armrest No
With Backrest Yes
View More Specs
Products 6
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S39211 Products for Science Education
View Documents
Black
Each for $195.00
Add to cart
S39210 Products for Science Education
View Documents
Black
Each for $195.00
Add to cart
S39208 Products for Science Education
View Documents
Black
Each for $195.00
Add to cart
S39209 Products for Science Education
View Documents
Black
Each for $195.00
Add to cart
S39357 Products for Science Education
View Documents
Black
Each for $195.00
Add to cart
S39213 Products for Science Education
View Documents
Black
Each for $195.00
Add to cart
Description
Description
  • Adjustable from 22-30 inches from the top of the seat to the floor
  • Features manual back and back depth adjustments
  • Black, powder-coated steel base with fixed foot ring
  • Stationary, non-marring glides suitable for hard floors
  • Antifungal/antibacterial Grade C Vinyl
Specifications
Specifications
Yes (Pneumatic)
No
Yes
No
Vinyl
No
No