Learn More
Honeywell CoreShield™ Cut Protection Gloves with 13 Gauge Liner
Feature a 13 Gauge Liner, and a Nitrile Coating. CoreShield™ Gloves ensure workers are not only protected but also comfortable and productive. A wide variety of additional key features enhance usability, wearability, and durability.
$11.50 - $13.00
Specifications
ANSI Abrasion Rating ANSI 6
For Use With (Application) Manufacturing, Automotive, Metal Stamping, Glass Handling, Construction, and Logistics
Product Type Cut Protection Gloves
Series CoreShield™
Style Slip-On
View More Specs
Products 6
Catalog Number Mfr. No. Cut Resistance Rating Size Price Quantity & Availability  
Catalog Number Mfr. No. Cut Resistance Rating Size Price Quantity & Availability  
19-085-709
View Documents
Honeywell
260913B/8M
ANSI A6 Medium Pair for $11.50
Case of 100 PR for $1,168.70
Add to cart
19-085-710
View Documents
Honeywell
260913B/9L
ANSI A6 Large Pair for $11.50
Case of 100 PR for $1,168.70
Add to cart
19-085-711
View Documents
Honeywell
260913B/10XL
ANSI A6 X-Large Pair for $11.50
Case of 100 PR for $1,168.70
Add to cart
19-085-712
View Documents
Honeywell
270513B/8M
ANSI A7 Medium Pair for $13.00
Case of 100 PR for $1,309.75
Add to cart
19-085-713
View Documents
Honeywell
270513B/9L
ANSI A7 Large Pair for $13.00
Case of 100 PR for $1,309.75
Add to cart
19-085-714
View Documents
Honeywell
270513B/10XL
ANSI A7 X-Large Pair for $13.00
Case of 100 PR for $1,309.75
Add to cart
Description
Description
  • CoreShield™ High Performance Coating with abrasion enhancement technology doubles glove durability
  • Specially designed elastic coating provides outstanding oil- and dry-grip performance
  • 360° breathability through an open cell structure provides enhanced comfort, for longer wear
  • Prevents hand fatigue for workers who need to wear gloves all day long
  • Longer cuff ensures wrist protection
  • Reinforced thumb for greater wear resistance and extended product life
  • Compatible with resistive touchscreen technology typically used on industrial machines and smartphones
  • No significant changes in performance or size even after 5 washing cycles 
  • Soft, skin-friendly yarns reduce allergy risk for wearers
Specifications
Specifications
ANSI 6
Cut Protection Gloves
Slip-On
Manufacturing, Automotive, Metal Stamping, Glass Handling, Construction, and Logistics
CoreShield™