Learn More
Honeywell CoreShield™ Cut Protection Gloves with 15 Gauge Liner
Features a Nitrile Liner, and a Nitrile Micro-Foam Coating. CoreShield™ Gloves ensure workers are not only protected but also comfortable and productive. A wide variety of additional key features enhance usability, wearability, and durability.
$3.60
Specifications
Coating Material Nitrile (Micro-Foam)
Color Black
Style Slip-On
Cut Resistance Rating ANSI A1
Hand Specific
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-085-655
View Documents
Honeywell
211515B/8M
Medium Pair for $3.60
Case of 100 PR for $360.68
Add to cart
19-085-726
View Documents
Honeywell
211515B/9L
Large Pair for $3.60
Case of 100 PR for $360.68
Add to cart
19-085-657
View Documents
Honeywell
211515B/10XL
X-Large Pair for $3.60
Case of 100 PR for $360.68
Add to cart
Description
Description
  • CoreShield™ High Performance Coating with abrasion enhancement technology doubles glove durability
  • Specially designed microfoam elastic coating provides outstanding oil- and dry-grip performance
  • 360° breathability through an open cell structure provides enhanced comfort, for longer wear
  • Prevents hand fatigue for workers who need to wear gloves all day long
  • Longer cuff ensures wrist protection
  • Reinforced thumb for greater wear resistance and extended product life
  • Compatible with resistive touchscreen technology typically used on industrial machines and smartphones
  • No significant changes in performance or size even after 5 washing cycles 
  • Soft, skin-friendly yarns reduce allergy risk for wearers
Specifications
Specifications
Nitrile (Micro-Foam)
Slip-On
Specific
Plastic Bag
Manufacturing, Automotive, Metal Stamping, Glass Handling, Construction, and Logistics
Reinforced
Black
ANSI A1
15 Gauge
CoreShield™
Cut Protection Gloves