Learn More
Honeywell CoreShield™ Cut Protection Gloves with ANSI A1 Compliance
Features an 18 Gauge Nitrile Liner, and a Nitrile Coating. CoreShield™ Gloves ensure workers are not only protected but also comfortable and productive. A wide variety of additional key features enhance usability, wearability, and durability.
$4.00
Specifications
Cut Resistance Rating ANSI A1
Lining 18 Gauge
Hand Specific
Color Black
Series CoreShield™
View More Specs
Products 6
Catalog Number Mfr. No. ANSI Abrasion Rating Size Price Quantity & Availability  
Catalog Number Mfr. No. ANSI Abrasion Rating Size Price Quantity & Availability  
19-085-661
View Documents
Honeywell
211818B/8M
ANSI 5 Medium Pair for $4.00
Case of 100 PR for $400.75
Add to cart
19-085-662
View Documents
Honeywell
211818B/9L
ANSI 5 Large Pair for $4.00
Case of 100 PR for $400.75
Add to cart
19-085-663
View Documents
Honeywell
211818B/10XL
ANSI 5 X-Large Pair for $4.00
Case of 100 PR for $400.75
Add to cart
19-085-658
View Documents
Honeywell
211518B/8M
ANSI 6 Medium Pair for $4.00
Case of 100 PR for $400.75
Add to cart
19-085-659
View Documents
Honeywell
211518B/9L
ANSI 6 Large Pair for $4.00
Case of 100 PR for $400.75
Add to cart
19-085-660
View Documents
Honeywell
211518B/10XL
ANSI 6 X-Large Pair for $4.00
Case of 100 PR for $400.75
Add to cart
Description
Description
  • CoreShield™ High Performance Coating with abrasion enhancement technology doubles glove durability
  • Specially designed elastic coating provides outstanding oil- and dry-grip performance
  • 360° breathability through an open cell structure provides enhanced comfort, for longer wear
  • Prevents hand fatigue for workers who need to wear gloves all day long
  • Longer cuff ensures wrist protection
  • Reinforced thumb for greater wear resistance and extended product life
  • Compatible with resistive touchscreen technology typically used on industrial machines and smartphones
  • No significant changes in performance or size even after 5 washing cycles 
  • Soft, skin-friendly yarns reduce allergy risk for wearers
Specifications
Specifications
ANSI A1
Specific
CoreShield™
Nylon
Cut Protection Gloves
Slip-On
18 Gauge
Black
Manufacturing, Automotive, Metal Stamping, Glass Handling, Construction, and Logistics
Plastic Bag
Reinforced