Learn More
Honeywell CoreShield™ Cut Protection Gloves with HPPE/Alloy/Basalt Liner
Features a 13 Gauge HPPE/Alloy/Basalt, and a Nitrile Coating. CoreShield™ Gloves ensure workers are not only protected but also comfortable and productive. A wide variety of additional key features enhance usability, wearability, and durability.
$10.50 - $11.50
Specifications
ANSI Abrasion Rating ANSI 6
Coating Material Nitrile (Micro-Foam)
Cut Resistance Rating ANSI A6
For Use With (Application) Manufacturing, Automotive, Metal Stamping, Glass Handling, Construction, and Logistics
Hand Specific
View More Specs
Products 6
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
19-085-703
View Documents
Honeywell
260513B/8M
Black Medium Pair for $11.50
Case of 100 PR for $1,168.70
Add to cart
19-085-704
View Documents
Honeywell
260513B/9L
Black Large Pair for $11.50
Case of 100 PR for $1,168.70
Add to cart
19-085-705
View Documents
Honeywell
260513B/10XL
Black X-Large Pair for $11.50
Case of 100 PR for $1,168.70
Add to cart
19-085-706
View Documents
Honeywell
260513W/8M
Gray Medium Pair for $10.50
Case of 100 PR for $1,057.88
Add to cart
19-085-707
View Documents
Honeywell
260513W/9L
Gray Large Pair for $10.50
Case of 100 PR for $1,057.88
Add to cart
19-085-708
View Documents
Honeywell
260513W/10XL
Gray X-Large Pair for $10.50
Case of 100 PR for $1,057.88
Add to cart
Description
Description
  • CoreShield™ High Performance Coating with abrasion enhancement technology doubles glove durability
  • Specially designed microfoam elastic coating provides outstanding oil- and dry-grip performance
  • 360° breathability through an open cell structure provides enhanced comfort, for longer wear
  • Prevents hand fatigue for workers who need to wear gloves all day long
  • Longer cuff ensures wrist protection
  • Reinforced thumb for greater wear resistance and extended product life
  • Compatible with resistive touchscreen technology typically used on industrial machines and smartphones
  • No significant changes in performance or size even after 5 washing cycles 
  • Soft, skin-friendly yarns reduce allergy risk for wearers
Specifications
Specifications
ANSI 6
ANSI A6
Specific
13 Gauge
CoreShield™
Nitrile (Micro-Foam)
Manufacturing, Automotive, Metal Stamping, Glass Handling, Construction, and Logistics
HPPE/Alloy/Basalt
Cut Protection Gloves
Slip-On