Learn More
Honeywell CoreShield™ Cut Protection Gloves with HPPE/Stainless Steel Lining
Features a 13 Gauge HPPE/Stainless Steel Liner, and a Nitrile Coating. CoreShield™ Gloves ensure workers are not only protected but also comfortable and productive. A wide variety of additional key features enhance usability, wearability, and durability.
$11.00 - $11.50
Specifications
ANSI Abrasion Rating ANSI 6
For Use With (Application) Manufacturing, Automotive, Metal Stamping, Glass Handling, Construction, and Logistics
Liner Material HDPE/Stainless Steel
Lining 13 Gauge
Product Type Cut Protection Gloves
View More Specs
Products 6
Catalog Number Mfr. No. Coating Material Size Price Quantity & Availability  
Catalog Number Mfr. No. Coating Material Size Price Quantity & Availability  
19-085-697
View Documents
Honeywell
250513B/8M
Nitrile (Micro-Foam) Medium Pair for $11.50
Case of 100 PR for $1,158.62
Add to cart
19-085-698
View Documents
Honeywell
250513B/9L
Nitrile (Micro-Foam) Large Pair for $11.50
Case of 100 PR for $1,158.62
Add to cart
19-085-699
View Documents
Honeywell
250513B/10XL
Nitrile (Micro-Foam) X-Large Pair for $11.50
Case of 100 PR for $1,158.62
Add to cart
19-085-700
View Documents
Honeywell
250913B/8M
Nitrile (Smooth) Medium Pair for $11.00
Case of 100 PR for $1,108.25
Add to cart
19-085-701
View Documents
Honeywell
250913B/9L
Nitrile (Smooth) Large Pair for $11.00
Case of 100 PR for $1,108.25
Add to cart
19-085-702
View Documents
Honeywell
250913B/10XL
Nitrile (Smooth) X-Large Pair for $11.00
Case of 100 PR for $1,108.25
Add to cart
Description
Description
  • CoreShield™ High Performance Coating with abrasion enhancement technology doubles glove durability
  • Specially designed elastic coating provides outstanding oil- and dry-grip performance
  • 360° breathability through an open cell structure provides enhanced comfort, for longer wear
  • Prevents hand fatigue for workers who need to wear gloves all day long
  • Longer cuff ensures wrist protection
  • Reinforced thumb for greater wear resistance and extended product life
  • Compatible with resistive touchscreen technology typically used on industrial machines and smartphones
  • No significant changes in performance or size even after 5 washing cycles 
  • Soft, skin-friendly yarns reduce allergy risk for wearers
Specifications
Specifications
ANSI 6
HDPE/Stainless Steel
Cut Protection Gloves
Slip-On
Manufacturing, Automotive, Metal Stamping, Glass Handling, Construction, and Logistics
13 Gauge
CoreShield™