Learn More
Honeywell CoreShield™ Cut Protection Gloves with Smooth Nitrile Coating
Feature a 13 Gauge Liner, and a Smooth Nitrile Coating. CoreShield™ Gloves ensure workers are not only protected but also comfortable and productive. A wide variety of additional key features enhance usability, wearability, and durability.
$7.50 - $10.00
Specifications
ANSI Abrasion Rating ANSI 6
For Use With (Application) Manufacturing, Automotive, Metal Stamping, Glass Handling, Construction, and Logistics
Lining 13 Gauge
Product Type Cut Protection Gloves
Series CoreShield™
View More Specs
Products 6
Catalog Number Mfr. No. Cut Resistance Rating Size Price Quantity & Availability  
Catalog Number Mfr. No. Cut Resistance Rating Size Price Quantity & Availability  
19-085-673
View Documents
Honeywell
227913B/8M
ANSI A2 Medium
Pair for $7.50
Case of 100 PR for $755.62
Add to cart
19-085-674
View Documents
Honeywell
227913B/9L
ANSI A2 Large
Pair for $7.50
Case of 100 PR for $755.62
Add to cart
19-085-675
View Documents
Honeywell
227913B/10XL
ANSI A2 X-Large
Pair for $7.50
Case of 100 PR for $755.62
Add to cart
19-085-691
View Documents
Honeywell
240913B/8M
ANSI A4 Medium
Pair for $10.00
Case of 100 PR for $1,007.50
Add to cart
19-085-692
View Documents
Honeywell
240913B/9L
ANSI A4 Large
Pair for $10.00
Case of 100 PR for $1,007.50
Add to cart
19-085-693
View Documents
Honeywell
240913B/10XL
ANSI A4 X-Large
Pair for $10.00
Case of 100 PR for $1,007.50
Add to cart
Description
Description
  • CoreShield™ High Performance Coating with abrasion enhancement technology doubles glove durability
  • Specially designed elastic coating provides outstanding oil- and dry-grip performance
  • 360° breathability through an open cell structure provides enhanced comfort, for longer wear
  • Prevents hand fatigue for workers who need to wear gloves all day long
  • Longer cuff ensures wrist protection
  • Reinforced thumb for greater wear resistance and extended product life
  • Compatible with resistive touchscreen technology typically used on industrial machines and smartphones
  • No significant changes in performance or size even after 5 washing cycles 
  • Soft, skin-friendly yarns reduce allergy risk for wearers
Specifications
Specifications
ANSI 6
13 Gauge
CoreShield™
Nitrile (Smooth)
Manufacturing, Automotive, Metal Stamping, Glass Handling, Construction, and Logistics
Cut Protection Gloves
Slip-On