Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CORIN Polyclonal Antibody
CORIN Polyclonal Antibody
Supplier: Thermo Scientific PA542916
Description
CORIN Polyclonal Antibody for Western Blot

Specifications
CORIN | |
Polyclonal | |
Unconjugated | |
CORIN | |
ATC2; Atrial natriuretic peptide-converting enzyme; Atrial natriuretic peptide-converting enzyme, 100 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, 160 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, 180 kDa soluble fragment; Atrial natriuretic peptide-converting enzyme, activated protease fragment; Atrial natriuretic peptide-converting enzyme, N-terminal propeptide; AV273130; Corin; corin, serine peptidase; CRN; EC 3.4.21; Heart-specific serine proteinase ATC2; low density lipoprotein receptor-related protein 4; low density lipoprotein-related protein 4; Lrp4; MGC119742; PEE5; pro-ANP-convertase; pro-ANP-converting enzyme; PRSC; serine peptidase; serine protease; TMPRSS10; Transmembrane protease serine 10 | |
Rabbit | |
Protein A | |
RUO | |
10699 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y5Q5 | |
CORIN | |
Synthetic peptide directed towards the C-terminal of human CORIN (aa 881-930). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction