Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corin Rat anti-Human, Alexa Fluor™ 594, Clone: MM0199-9H21, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212205AF594

Catalog No. NP21220A94


Add to cart

Description

Description

Corin Monoclonal antibody specifically detects Corin in Human samples. It is validated for Western Blot.
Specifications

Specifications

Corin
MM0199-9H21
Western Blot
atrial natriuteric peptide-converting enzyme, Corin, corin, serine peptidase, EC 3.4.21, EC 3.4.21.-, heart specific serine proteinase, Heart-specific serine proteinase ATC2, MGC119742, pro-ANP-convertase, Pro-ANP-converting enzyme, PRSC, serine protease, Transmembrane protease serine 10
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CORIN
Human Corin
0.1 mL
Primary
10699
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit