Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corn Oil, NF, Spectrum™

Click to view available options
Quantity:
20 L
200 L
25 mL
4 L
500 mL
Packaging:
Amber Glass Bottle
Drum
Steel Can
8001-30-7
8001-30-7

CO136, 8001-30-7

Spectrum™ Corn Oil, NF its uses include as a solvent vehicle for topical preparations and as a glidant excipient for oral dosage tablets and capsules. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 4 L
CAS Max % 100%