Learn More
Corn Oil, NF, Spectrum™
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
8001-30-7
8001-30-7
CO136, 8001-30-7
Spectrum™ Corn Oil, NF its uses include as a solvent vehicle for topical preparations and as a glidant excipient for oral dosage tablets and capsules. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Steel Can
Quantity 20 L
CAS Max % 1