Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corn Starch, Powder, NF, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
9005-25-8
C12H22O11
342.30
GUBGYTABKSRVRQ-UHFFFAOYNA-N
2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
9005-25-8
C12H22O11
342.30
GUBGYTABKSRVRQ-UHFFFAOYNA-N
2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

S1552, 9005-25-8, (C6H10O5)n

Spectrum™ Corn Starch, Powder, NF is used as filler, binder and disintegrant excipient. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Ignition Residue 0.6%
Loss on Drying 15%
Packaging Poly Bottle
Quantity 500 g