Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Corning™ 384-Well Low-Volume Solid Microplates
Same low reagent volumes as 1536-well plates without the need for new instrumentation
Manufacturer:  Corning™ 3676
 View more versions of this product
Catalog No. 12-595-460
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Black
384-well
384
Tissue Culture
2 to 20μL
Round
Provides easier access and imaging for superior assay performance, fluorescence applications, reduce cross-contamination during assay and liquid-handling process
Polystyrene
No
50μL
Black