Corning™ 384-Well Low-Volume Solid Microplates
Same low reagent volumes as 1536-well plates without the need for new instrumentation
Manufacturer: Corning™ 3677
 View more versions of this product
Catalog No. 12-595-461
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Color Black
    For Use With (Application) Provides easier access and imaging for superior assay performance, fluorescence applications, reduce cross-contamination during assay and liquid-handling process
    Format 384-well
    Material Polystyrene
    No. of Wells 384
    Sterile No
    Surface Coating Non-treated
    Volume (Metric) Well 35μL
    Volume (Metric) Working 2 to 25μL
    Well Color Black
    Well Design Round
    Polystyrene plates feature a unique square-to-round well design with a conical-shaped bottom that provides easier access and imaging for superior assay performance.
    • Bottom reduces fluid and air entrapment, greater signal-to-noise ratio and reduced FP signal (mP) when working with very low volumes
    • Each well has a raised rim to reduce cross-contamination during assay and liquid-handling process
    • Choose black plates for fluorescence applications or white for luminescence applications
    High-Binding Surface Plates
    • Ideal for immunoassays
    • Can bind medium (>10kDa) and large biomolecules that possess ionic groups and/or hydrophobic regions
    • Working volume: 2 to 20µL
  • Description & Specifications
Related Products
Product Content Feedback