Corning™ 384-Well Low-Volume Solid Microplates
Same low reagent volumes as 1536-well plates without the need for new instrumentation
Manufacturer: Corning™ 3673
 View more versions of this product
Catalog No. 07-200-330
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    For Use With (Application) Provides easier access and imaging for superior assay performance, fluorescence applications, reduce cross-contamination during assay and liquid-handling process
    Format 384-well
    Material Polystyrene
    No. of Wells 384
    Sterile No
    Surface Coating NBS™
    Volume (Metric) Well 50μL
    Volume (Metric) Working 5 to 40μL
    Well Color White,White
    Well Design Round
    Color White
    Applications Provides easier access and imaging for superior assay performance, fluorescence applications, reduce cross-contamination during assay and liquid-handling process
    Well Shape Round
    Wells 384
  • Description & Specifications
Related Products
Product Content Feedback