Corning™ 384-Well Nonbinding Surface (NBS™) Microplates Available on GSA/VA Contract for Federal Government customers only.

Enhanced signal-to-noise ratio

Manufacturer: Corning™ 3653BC

 View more versions of this product

Catalog No. 07-200-870

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Application) For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
    For Use With (Equipment) Plate readers, liquid handlers and robotic systems
    Format 384-well
    Sterile No
    Surface Coating NBS™
    Temperature Range (English) 39° to 99°F
    Temperature Range (Metric) 4° to 37°C
    Volume (Metric) Well 112μL,112μL
    Volume (Metric) Working 20 to 80μL
    Well Color Clear,Clear
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit