Learn More
Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates permit top or bottom analysis using detection microplate readers.
Manufacturer:  Corning™ 3712
 View more versions of this product
Catalog No. 07-200-655
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Molded polystyrene with opaque walls and optically clear bottoms
  • Readable from top or bottom for direct microscope viewing and CCD camera-based detectors
  • Opaque black or white walls prevent well to well crosstalk
  • Recommended working volumes of 20 to 80µL; total well volume of 112µL
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Suited for flourescent and luminescent assays
  • Plate dimensions meet proposed industry standard
Specifications
Specifications
No
Flat
0.06cm2
384 well
384
20 to 80μL
With Lid
384-Well TC-treated Microplate
Tissue-Culture Treated
Black
Attachment and growth of anchorage-dependent cells
Polystyrene
112μL
Clear
112μL
Sterile