Learn More
Corning™ 384-Well Solid Black or White Polystyrene Microplates Available on GSA/VA Contract for Federal Government customers only.
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Manufacturer:  Corning™ 3826BC
 View more versions of this product
Catalog No. 07-201-13
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Minimal well to well crosstalk
  • White microplates enhance luminecent signals and have low background luminescence
  • Black mircoplates have low background flourescence and minimize light scattering
  • Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
  • Medical-grade polystyrene
  • Well volume: 112µL
  • Working volume: 20µL to 80µL
  • Surface area/well: 0.06cm2
  • Treated surface options are not coated, require no special handling and are stable at room temperature
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Plate dimensions meet proposed industry standards
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Tissue-Culture Treated
White
Attachment and growth of anchorage-dependent cells
Generic barcodes
384
5 to 40μL
With Lid
384-Well Low Volume Microplate
Flat
0.03cm2
Low volume
Polystyrene
50μL
White
50μL
Sterile